Assay Details
| Gene Symbol: mtus1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077608.2 | NP_001071076.2 | 9 - 10 | 1233 | 88 | |
| XM_005157240.4 | XP_005157297.1 | 14 - 15 | 3521 | 88 | ||
| XM_005157242.4 | XP_005157299.1 | 8 - 9 | 928 | 88 | ||
| XM_017359019.2 | XP_017214508.1 | 14 - 15 | 3518 | 88 | ||
| XM_021481039.1 | XP_021336714.1 | 11 - 12 | 1468 | 88 | ||
| GenBank mRNA | BC125970.1 | - | 9 - 10 | 1233 | 88 | |
More Information
Set Membership: |
|

mtus1a
Genomic Map