Assay Details
| Gene Symbol: dars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077611.1 | NP_001071079.1 | 13 - 14 | 1235 | 61 | |
| GenBank mRNA | BC124800.1 | - | 13 - 14 | 1235 | 61 | |
More Information
Set Membership: |
|

dars1
Genomic Map