Assay Details
| Gene Symbol: bida | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079826.1 | NP_001073295.1 | 1 - 2 | 258 | 82 | |
| XM_005159109.4 | XP_005159166.1 | 2 - 3 | 383 | 82 | ||
| GenBank mRNA | BC162699.1 | - | 1 - 2 | 307 | 82 | |
| BC162712.1 | - | 1 - 2 | 307 | 82 | ||
| DQ860155.1 | - | 1 - 2 | 258 | 82 | ||
More Information
Set Membership: |
|

bida
Genomic Map