Assay Details
| Gene Symbol: bida | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079826.1 | NP_001073295.1 | 2 - 3 | 393 | 91 | |
| XM_005159109.4 | XP_005159166.1 | 3 - 4 | 518 | 91 | ||
| GenBank mRNA | BC162699.1 | - | 2 - 3 | 442 | 91 | |
| BC162712.1 | - | 2 - 3 | 442 | 91 | ||
| DQ860155.1 | - | 2 - 3 | 91 | |||
More Information
Set Membership: |
|

bida
Genomic Map