Assay Details
| Gene Symbol: nt5c2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079966.2 | NP_001073435.2 | 12 - 13 | 1121 | 67 | |
| XM_005168392.4 | XP_005168449.1 | 12 - 13 | 1290 | 67 | ||
| XM_021474992.1 | XP_021330667.1 | 11 - 12 | 965 | 67 | ||
| XM_021474993.1 | XP_021330668.1 | 7 - 8 | 483 | 67 | ||
| GenBank mRNA | BC076085.1 | - | 12 - 13 | 1220 | 67 | |
| BC095068.1 | - | 12 - 13 | 1372 | 67 | ||
| BC124713.1 | - | 12 - 13 | 1066 | 67 | ||
More Information
Set Membership: |
|

nt5c2b
Genomic Map