Assay Details
| Gene Symbol: nt5c2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079966.2 | NP_001073435.2 | 14 - 15 | 1364 | 66 | |
| XM_005168392.4 | XP_005168449.1 | 14 - 15 | 1533 | 66 | ||
| XM_021474992.1 | XP_021330667.1 | 13 - 14 | 1208 | 66 | ||
| XM_021474993.1 | XP_021330668.1 | 9 - 10 | 726 | 66 | ||
| GenBank mRNA | BC076085.1 | - | 14 - 15 | 1463 | 66 | |
| BC095068.1 | - | 14 - 15 | 1615 | 66 | ||
| BC124713.1 | - | 14 - 15 | 1309 | 66 | ||
More Information
Set Membership: |
|

nt5c2b
Genomic Map