Assay Details
| Gene Symbol: ano5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079983.1 | NP_001073452.1 | 16 - 17 | 2392 | 78 | |
| XM_005168891.4 | XP_005168948.1 | 17 - 18 | 2529 | 78 | ||
| XM_009303225.3 | XP_009301500.1 | 15 - 16 | 2195 | 78 | ||
| GenBank mRNA | BC128870.1 | - | 16 - 17 | 2392 | 78 | |
More Information
Set Membership: |
|

ano5b
Genomic Map