Assay Details
| Gene Symbol: sirt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080174.1 | NP_001073643.1 | 2 - 3 | 254 | 69 | |
| XM_009303487.3 | XP_009301762.1 | 2 - 3 | 289 | 69 | ||
| XM_009303488.3 | XP_009301763.1 | 2 - 3 | 323 | 69 | ||
| XM_017356877.2 | XP_017212366.1 | 2 - 3 | 289 | 69 | ||
| GenBank mRNA | BC129442.1 | - | 2 - 3 | 254 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009303488.3 transcripts.
Set Membership: |
|

sirt3
Genomic Map