Assay Details
| Gene Symbol: ptpmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080187.1 | NP_001073656.1 | 3 - 4 | 526 | 88 | |
| GenBank mRNA | BC129407.1 | - | 3 - 4 | 526 | 88 | |
More Information
Set Membership: |
|

ptpmt1
Genomic Map