Assay Details
| Gene Symbol: mavs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080584.2 | NP_001074053.2 | 5 - 6 | 1101 | 90 | |
| GenBank mRNA | BC129221.1 | - | 5 - 6 | 1060 | 90 | |
More Information
Set Membership: |
|

mavs
Genomic Map