Assay Details
| Gene Symbol: mfge8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080990.1 | NP_001074459.1 | 6 - 7 | 845 | 90 | |
| XM_005163016.4 | XP_005163073.1 | 6 - 7 | 910 | 90 | ||
| GenBank mRNA | BC131837.1 | - | 6 - 7 | 845 | 90 | |
More Information
Set Membership: |
|

mfge8b
Genomic Map