Assay Details
| Gene Symbol: mfge8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080990.1 | NP_001074459.1 | 7 - 8 | 995 | 66 | |
| GenBank mRNA | BC131837.1 | - | 7 - 8 | 995 | 66 | |
More Information
Set Membership: |
|

mfge8b
Genomic Map