Assay Details
| Gene Symbol: tbl1x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082994.1 | NP_001076463.1 | 12 - 13 | 1500 | 75 | |
| GenBank mRNA | BC133078.1 | - | 12 - 13 | 1500 | 75 | |
More Information
Set Membership: |
|

tbl1x
Genomic Map