Assay Details
| Gene Symbol: sphkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083045.1 | NP_001076514.1 | 5 - 6 | 513 | 72 | |
| XM_009293856.3 | XP_009292131.1 | 6 - 7 | 891 | 72 | ||
| XM_009293857.3 | XP_009292132.1 | 5 - 6 | 756 | 72 | ||
| XM_009293859.3 | XP_009292134.1 | 6 - 7 | 889 | 72 | ||
More Information
Set Membership: |
|

sphkap
Genomic Map