Assay Details
| Gene Symbol: tnksb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089415.2 | NP_001082884.1 | 21 - 22 | 3137 | 108 | |
| XM_005171743.4 | XP_005171800.1 | 22 - 23 | 3201 | 108 | ||
| XM_005171744.4 | XP_005171801.1 | 21 - 22 | 3180 | 108 | ||
| XM_005171745.4 | XP_005171802.1 | 22 - 23 | 3170 | 108 | ||
| GenBank mRNA | BC162262.1 | - | 21 - 22 | 3137 | 108 | |
| BC162268.1 | - | 21 - 22 | 3137 | 108 | ||
| Gene Symbol: tnksa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CK028164.1 | - | 3 - 4 | 323 | 108 | |
More Information
Set Membership: |
|

tnksb
Genomic Map