Assay Details
| Gene Symbol: tnksb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089415.2 | NP_001082884.1 | 25 - 26 | 3611 | 70 | |
| XM_005171743.4 | XP_005171800.1 | 26 - 27 | 3675 | 70 | ||
| XM_005171744.4 | XP_005171801.1 | 25 - 26 | 3654 | 70 | ||
| XM_005171745.4 | XP_005171802.1 | 26 - 27 | 3644 | 70 | ||
| GenBank mRNA | BC162262.1 | - | 25 - 26 | 3611 | 70 | |
| BC162268.1 | - | 25 - 26 | 3611 | 70 | ||
More Information
Set Membership: |
|

tnksb
Genomic Map