Assay Details
| Gene Symbol: zgc:198329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098246.2 | NP_001091716.1 | 7 - 8 | 1155 | 93 | |
| GenBank mRNA | BC171602.1 | - | 7 - 8 | 1108 | 93 | |
| BC171606.1 | - | 7 - 8 | 1108 | 93 | ||
| CU458935.1 | - | 7 - 8 | 1123 | 93 | ||
More Information
Set Membership: |
|

zgc:198329
Genomic Map