Assay Details
| Gene Symbol: atp5meb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098481.1 | NP_001091951.1 | 2 - 3 | 159 | 131 | |
| GenBank mRNA | BC124416.1 | - | 2 - 3 | 159 | 131 | |
More Information
Set Membership: |
|

atp5meb
Genomic Map