Assay Details
| Gene Symbol: poglut3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098745.1 | NP_001092215.1 | 4 - 5 | 1142 | 134 | |
| GenBank mRNA | BC142839.1 | - | 4 - 5 | 1142 | 134 | |
More Information
Set Membership: |
|

poglut3
Genomic Map