Assay Details
| Gene Symbol: cemip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099449.1 | NP_001092919.1 | 12 - 13 | 2405 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cemip2| Gene Symbol: cemip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099449.1 | NP_001092919.1 | 12 - 13 | 2405 | 65 | |
Set Membership: |
|