Assay Details
| Gene Symbol: cemip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099449.1 | NP_001092919.1 | 14 - 15 | 2746 | 138 | |
| XM_005171900.4 | XP_005171957.1 | 16 - 17 | 3020 | 138 | ||
| GenBank mRNA | HQ997922.1 | - | 15 - 16 | 2782 | 138 | |
More Information
Set Membership: |
|

cemip2
Genomic Map