Assay Details
| Gene Symbol: cemip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099449.1 | NP_001092919.1 | 9 - 10 | 2056 | 81 | |
| XM_005171900.4 | XP_005171957.1 | 10 - 11 | 2294 | 81 | ||
| GenBank mRNA | HQ997922.1 | - | 9 - 10 | 2056 | 81 | |
More Information
Set Membership: |
|

cemip2
Genomic Map