Assay Details
| Gene Symbol: spag9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021470835.1 | XP_021326510.1 | 5 - 6 | 1306 | 113 | |
| GenBank mRNA | BC150225.1 | - | 5 - 6 | 907 | 113 | |
More Information
Set Membership: |
|

spag9a
Genomic Map