Assay Details
| Gene Symbol: spag9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320288.1 | NP_001307217.1 | 8 - 9 | 1250 | 70 | |
| XM_017354369.2 | XP_017209858.1 | 9 - 10 | 1664 | 70 | ||
| XM_017354374.2 | XP_017209863.1 | 9 - 10 | 1664 | 70 | ||
| XM_021470820.1 | XP_021326495.1 | 9 - 10 | 1664 | 70 | ||
| XM_021470831.1 | XP_021326506.1 | 8 - 9 | 1649 | 70 | ||
| XM_021470832.1 | XP_021326507.1 | 9 - 10 | 1664 | 70 | ||
| XM_021470833.1 | XP_021326508.1 | 9 - 10 | 1664 | 70 | ||
| XM_021470835.1 | XP_021326510.1 | 8 - 9 | 1583 | 70 | ||
| GenBank mRNA | BC150225.1 | - | 6 - 7 | 1594 | 70 | |
More Information
Set Membership: |
|

spag9a
Genomic Map