Assay Details
| Gene Symbol: zdhhc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110026.1 | NP_001103496.1 | 1 - 2 | 163 | 59 | |
| GenBank mRNA | BC115336.1 | - | 1 - 2 | 163 | 59 | |
More Information
Set Membership: |
|

zdhhc9
Genomic Map