Assay Details
| Gene Symbol: calcoco1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110166.1 | NP_001103636.1 | 7 - 8 | 1070 | 112 | |
| XM_009297039.3 | XP_009295314.1 | 8 - 9 | 1105 | 112 | ||
More Information
Set Membership: |
|

calcoco1a
Genomic Map