Assay Details
| Gene Symbol: calcoco1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110166.1 | NP_001103636.1 | 10 - 11 | 1604 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

calcoco1a| Gene Symbol: calcoco1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110166.1 | NP_001103636.1 | 10 - 11 | 1604 | 89 | |
Set Membership: |
|