Assay Details
| Gene Symbol: serpinh1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110374.2 | NP_001103844.1 | 3 - 4 | 836 | 68 | |
| GenBank mRNA | BC154462.1 | - | 3 - 4 | 843 | 68 | |
More Information
Set Membership: |
|

serpinh1a
Genomic Map