Assay Details
| Gene Symbol: tmx2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110387.1 | NP_001103857.1 | 5 - 6 | 571 | 131 | |
| XM_017359018.2 | XP_017214507.1 | 3 - 4 | 295 | 131 | ||
| GenBank mRNA | BC122455.1 | - | 5 - 6 | 594 | 131 | |
| BC154620.1 | - | 5 - 6 | 571 | 131 | ||
More Information
Set Membership: |
|

tmx2a
Genomic Map