Assay Details
| Gene Symbol: tmx2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110387.1 | NP_001103857.1 | 6 - 7 | 631 | 80 | |
| XM_017359018.2 | XP_017214507.1 | 4 - 5 | 355 | 80 | ||
| GenBank mRNA | BC122455.1 | - | 6 - 7 | 654 | 80 | |
| BC154620.1 | - | 6 - 7 | 631 | 80 | ||
More Information
Set Membership: |
|

tmx2a
Genomic Map