Assay Details
| Gene Symbol: tmx2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110387.1 | NP_001103857.1 | 7 - 8 | 767 | 124 | |
| GenBank mRNA | BC154620.1 | - | 7 - 8 | 767 | 124 | |
More Information
Set Membership: |
|

tmx2a
Genomic Map