Assay Details
| Gene Symbol: mapk8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110389.1 | NP_001103859.1 | 10 - 11 | 1176 | 101 | |
| XM_005156857.4 | XP_005156914.1 | 11 - 12 | 1399 | 101 | ||
| XM_005156858.4 | XP_005156915.1 | 11 - 12 | 1398 | 101 | ||
| XM_021480607.1 | XP_021336282.1 | 11 - 12 | 1285 | 101 | ||
| GenBank mRNA | BC154588.1 | - | 10 - 11 | 1176 | 101 | |
More Information
Set Membership: |
|

mapk8a
Genomic Map