Assay Details
| Gene Symbol: mapk8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110389.1 | NP_001103859.1 | 7 - 8 | 904 | 80 | |
| XM_005156857.4 | XP_005156914.1 | 8 - 9 | 1127 | 80 | ||
| XM_005156858.4 | XP_005156915.1 | 8 - 9 | 1126 | 80 | ||
| XM_017358755.2 | XP_017214244.1 | 8 - 9 | 1128 | 80 | ||
| XM_017358756.2 | XP_017214245.1 | 8 - 9 | 1128 | 80 | ||
| XM_021480607.1 | XP_021336282.1 | 8 - 9 | 1013 | 80 | ||
| GenBank mRNA | BC154588.1 | - | 7 - 8 | 904 | 80 | |
More Information
Set Membership: |
|

mapk8a
Genomic Map