Assay Details
| Gene Symbol: lfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130971.1 | NP_571046.1 | 1 - 2 | 546 | 99 | |
| GenBank mRNA | AY007434.1 | - | 1 - 2 | 546 | 99 | |
| BC044339.1 | - | 1 - 2 | 523 | 99 | ||
| BC164773.1 | - | 1 - 2 | 444 | 99 | ||
More Information
Set Membership: |
|

lfng
Genomic Map