Assay Details
| Gene Symbol: cmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131014.1 | NP_571089.1 | 17 - 18 | 895 | 63 | |
| XM_005156049.4 | XP_005156106.1 | 17 - 18 | 971 | 63 | ||
| XM_005156050.4 | XP_005156107.1 | 16 - 17 | 928 | 63 | ||
| XM_009306481.3 | XP_009304756.1 | 17 - 18 | 971 | 63 | ||
| XM_009306483.3 | XP_009304758.1 | 9 - 10 | 563 | 63 | ||
| XM_017358425.2 | XP_017213914.1 | 17 - 18 | 971 | 63 | ||
| XM_017358426.2 | XP_017213915.1 | 14 - 15 | 818 | 63 | ||
| XM_021480020.1 | XP_021335695.1 | 17 - 18 | 971 | 63 | ||
| GenBank mRNA | AF102865.1 | - | 17 - 18 | 895 | 63 | |
| BC090694.1 | - | 17 - 18 | 944 | 63 | ||
More Information
Set Membership: |
|

cmn
Genomic Map