Assay Details
| Gene Symbol: cmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131014.1 | NP_571089.1 | 18 - 19 | 948 | 84 | |
| XM_005156049.4 | XP_005156106.1 | 18 - 19 | 1024 | 84 | ||
| XM_005156050.4 | XP_005156107.1 | 17 - 18 | 981 | 84 | ||
| XM_009306481.3 | XP_009304756.1 | 18 - 19 | 1024 | 84 | ||
| XM_009306483.3 | XP_009304758.1 | 10 - 11 | 616 | 84 | ||
| XM_017358425.2 | XP_017213914.1 | 18 - 19 | 1024 | 84 | ||
| XM_017358426.2 | XP_017213915.1 | 15 - 16 | 871 | 84 | ||
| XM_021480020.1 | XP_021335695.1 | 18 - 19 | 1024 | 84 | ||
| GenBank mRNA | AF102865.1 | - | 18 - 19 | 948 | 84 | |
| BC090694.1 | - | 18 - 19 | 997 | 84 | ||
More Information
Set Membership: |
|

cmn
Genomic Map