Assay Details
| Gene Symbol: cmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131014.1 | NP_571089.1 | 33 - 34 | 1718 | 83 | |
| GenBank mRNA | AF102865.1 | - | 33 - 34 | 1718 | 83 | |
| BC090694.1 | - | 33 - 34 | 1767 | 83 | ||
More Information
Set Membership: |
|

cmn
Genomic Map