Assay Details
| Gene Symbol: cmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131014.1 | NP_571089.1 | 46 - 47 | 3025 | 62 | |
| XM_005156049.4 | XP_005156106.1 | 48 - 49 | 3227 | 62 | ||
| XM_005156050.4 | XP_005156107.1 | 47 - 48 | 3184 | 62 | ||
| XM_009306481.3 | XP_009304756.1 | 43 - 44 | 2972 | 62 | ||
| XM_009306483.3 | XP_009304758.1 | 40 - 41 | 2819 | 62 | ||
| XM_009306485.3 | XP_009304760.1 | 37 - 38 | 2666 | 62 | ||
| XM_017358425.2 | XP_017213914.1 | 46 - 47 | 3125 | 62 | ||
| XM_017358426.2 | XP_017213915.1 | 45 - 46 | 3074 | 62 | ||
| XM_021480020.1 | XP_021335695.1 | 45 - 46 | 3074 | 62 | ||
| XM_021480021.1 | XP_021335696.1 | 37 - 38 | 2666 | 62 | ||
| GenBank mRNA | AF102865.1 | - | 46 - 47 | 3025 | 62 | |
More Information
Set Membership: |
|

cmn
Genomic Map