Assay Details
| Gene Symbol: fsta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131037.3 | NP_571112.3 | 4 - 5 | 803 | 104 | |
| XM_005165257.4 | XP_005165314.1 | 4 - 5 | 863 | 104 | ||
| XM_009301618.3 | XP_009299893.1 | 4 - 5 | 869 | 104 | ||
| GenBank mRNA | AF084948.1 | - | 4 - 5 | 781 | 104 | |
| BC078241.1 | - | 4 - 5 | 804 | 104 | ||
| BC152095.1 | - | 4 - 5 | 797 | 104 | ||
| DQ317968.1 | - | 4 - 5 | 769 | 104 | ||
| JN662961.1 | - | 2 - 3 | 258 | 104 | ||
| KM655617.1 | - | 4 - 5 | 728 | 104 | ||
More Information
Set Membership: |
|

fsta
Genomic Map