Assay Details
| Gene Symbol: sema3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131050.2 | NP_571125.2 | 13 - 14 | 1553 | 108 | |
| GenBank mRNA | AF073290.1 | - | 3 - 4 | 216 | 108 | |
| AY860954.1 | - | 13 - 14 | 1475 | 108 | ||
| CU638804.1 | - | 13 - 14 | 1475 | 108 | ||
More Information
Set Membership: |
|

sema3h
Genomic Map