Assay Details
Gene Symbol: kita | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131053.1 | NP_571128.1 | 2 - 3 | 376 | 72 | |
GenBank mRNA | AF153446.1 | - | 2 - 3 | 376 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
