Assay Details
| Gene Symbol: kita | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131053.1 | NP_571128.1 | 3 - 4 | 664 | 84 | |
| XM_009294634.3 | XP_009292909.1 | 3 - 4 | 709 | 84 | ||
| XM_009294635.3 | XP_009292910.1 | 3 - 4 | 739 | 84 | ||
| GenBank mRNA | AF153446.1 | - | 3 - 4 | 664 | 84 | |
| BC162502.1 | - | 3 - 4 | 679 | 84 | ||
| CU458898.1 | - | 3 - 4 | 615 | 84 | ||
More Information
Set Membership: |
|

kita
Genomic Map