Assay Details
| Gene Symbol: kita | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131053.1 | NP_571128.1 | 7 - 8 | 1274 | 66 | |
| XM_009294634.3 | XP_009292909.1 | 7 - 8 | 1319 | 66 | ||
| XM_009294635.3 | XP_009292910.1 | 7 - 8 | 1349 | 66 | ||
| GenBank mRNA | AF153446.1 | - | 7 - 8 | 1274 | 66 | |
| BC162502.1 | - | 7 - 8 | 1289 | 66 | ||
| CU458898.1 | - | 7 - 8 | 1225 | 66 | ||
More Information
Set Membership: |
|

kita
Genomic Map