Assay Details
| Gene Symbol: rxrga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131217.4 | NP_571292.3 | 2 - 3 | 338 | 61 | |
| GenBank mRNA | BC059576.1 | - | 2 - 3 | 309 | 61 | |
| EH435141.1 | - | 2 - 3 | 366 | 61 | ||
| U29940.1 | - | 2 - 3 | 260 | 61 | ||
More Information
Set Membership: |
|

rxrga
Genomic Map