Assay Details
| Gene Symbol: gsk3ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131381.1 | NP_571456.1 | 6 - 7 | 1418 | 77 | |
| XM_009304494.3 | XP_009302769.1 | 6 - 7 | 1498 | 77 | ||
| GenBank mRNA | AB032265.1 | - | 6 - 7 | 954 | 77 | |
| AJ223502.1 | - | 6 - 7 | 1418 | 77 | ||
| BC162371.1 | - | 6 - 7 | 1351 | 77 | ||
More Information
Set Membership: |
|

gsk3ba
Genomic Map