Assay Details
| Gene Symbol: ephb4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131414.1 | NP_571489.1 | 8 - 9 | 1702 | 97 | |
| GenBank mRNA | AJ005026.2 | - | 8 - 9 | 1702 | 97 | |
More Information
Set Membership: |
|

ephb4a
Genomic Map