Assay Details
| Gene Symbol: ephb4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131414.1 | NP_571489.1 | 9 - 10 | 1803 | 78 | |
| XM_021476001.1 | XP_021331676.1 | 9 - 10 | 2568 | 78 | ||
| GenBank mRNA | AJ005026.2 | - | 9 - 10 | 1803 | 78 | |
More Information
Set Membership: |
|

ephb4a
Genomic Map