Assay Details
| Gene Symbol: fn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131520.2 | NP_571595.2 | 16 - 17 | 2550 | 75 | |
| XM_009304503.3 | XP_009302778.1 | 16 - 17 | 2587 | 75 | ||
| GenBank mRNA | AF081127.1 | - | 16 - 17 | 2549 | 75 | |
| AF081128.1 | - | 16 - 17 | 2502 | 75 | ||
| BC162574.1 | - | 16 - 17 | 2489 | 75 | ||
More Information
Set Membership: |
|

fn1a
Genomic Map