Assay Details
| Gene Symbol: fn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131520.2 | NP_571595.2 | 17 - 18 | 2642 | 80 | |
| XM_009304503.3 | XP_009302778.1 | 17 - 18 | 2679 | 80 | ||
| GenBank mRNA | AF081127.1 | - | 17 - 18 | 2641 | 80 | |
| AF081128.1 | - | 17 - 18 | 2594 | 80 | ||
| BC162574.1 | - | 17 - 18 | 2581 | 80 | ||
More Information
Set Membership: |
|

fn1a
Genomic Map