Assay Details
| Gene Symbol: fn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131520.2 | NP_571595.2 | 26 - 27 | 4392 | 75 | |
| XM_009304503.3 | XP_009302778.1 | 26 - 27 | 4429 | 75 | ||
| GenBank mRNA | AF081128.1 | - | 26 - 27 | 4341 | 75 | |
| BC162574.1 | - | 26 - 27 | 4331 | 75 | ||
More Information
Set Membership: |
|

fn1a
Genomic Map